Would you like to log in again?
Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
710107 768160-TRI 607107 606160 710160 710180 760107 607180 606180 604160-TRI 710183 604107 604181 710160-TRI 607160-TRI 604160 760160-TRI 607160 768180 760160 768160 606107 606160-TRI 760180
82051-584CS 584.55 USD InStock
82051-584 75799-852 82050-070 75799-862 82051-182 75799-864 89085-300 82050-072 82051-118 82051-116 75799-848 82051-212 82050-478 82050-006 82050-480 82050-074 82050-482 75799-850 82050-472 75799-860 82050-000 82050-474 82050-002 82050-476
Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Pipettes Serological Pipettes
Pipettes are manufactured from optical-grade polystyrene and feature a high internal and external surface polish, which reduces meniscus formation and helps ensure complete liquid expulsion.
  • Polystyrene construction
  • Polish reduces meniscus formation
  • Free of RNase, DNase, and pyrogens
  • Designed to fit automatic, semiautomatic, and manual pipetting devices
Fine-line graduation printing further contributes to accuracy of at least ±2%. Each disposable pipette is cotton-plugged and color-coded to facilitate identification. Pipettes with ascending and descending graduations allow easy reading of both aspirated and dispensing volumes. Most pipettes feature an improved anti-drip tip design. All pipettes are RNase-free, DNase-free, pyrogen-free, and noncytotoxic.
Ordering information: The 2 ml aspirating pipette that has no graduations or color band is supplied without a cotton plug. Individually wrapped items provide reduced contamination risk and particulate-free opening. Paper/plastic packaging offers both peel-off and break-through access. Plastic/plastic packaging offers peel-off access for easy opening and aseptic removal. Printed information on each individual package provides catalog number, product description, lot number, and best-before date. Durable bulk packaged items help eliminate bag puncture and a ziplock closure helps maintain product sterility.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty