Would you like to log in again?
Semi-Micro/Macro Cuvette, Polystyrene, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
613101 614101
82050-500CS 161.19 USD InStock
82050-500 82050-502
Semi-Micro/Macro Cuvette, Polystyrene, Greiner Bio-One
Cuvettes Spectrophotometer Cuvettes
Ideal for enzymatic determinations, since the very thin walls enable rapid and uniform equilibration.
  • Polystyrene construction
  • Minimal light scatter
  • High transmission rate
  • Very thin walls
Characterized by minimal light scatter but high transmission rate.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty