Would you like to log in again?
Disposal Bags, Polypropylene, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
646401 649401 649201 643201 646201
82050-610CS 372.6 USD InStock
82050-610 82050-612 82050-602 82050-616 10859-854
Disposal Bags, Polypropylene, Greiner Bio-One
Bags Waste Disposal Bags
Greiner Bio -One disposal bags are ideal for the disposal of contaminated laboratory items.
  • Autoclavable
  • Polypropylene construction
  • Disposal bags may be marked with 'biohazard' upon request
Standard bags are made of polypropylene foil for sterilization in the steam autoclave.
Also available upon request with imprint 'biohazard'.
Oxygen transport rate of 50 µm PP foil.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty