Would you like to log in again?
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
Supplier: Greiner Bio-One
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
650001 650101 651061 651101 650061 655081 655061-25 651001 655080 655001 655085 655101 655061
89136-728CS 138.95 USD InStock
89136-728 82050-716 82050-618 82050-620 82050-720 82050-622 82050-734 89041-276 89130-320 82050-650 82050-760 82050-652 82050-740
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
Microplates HTS Microplates
These 96-well microplates are available with flat-, round-, conical-, and C-bottom well styles, and are manufactured from clear, virgin polystyrene and selected for optical clarity and consistency.
  • Clear virgin polystyrene construction
  • High degree of optical clarity
  • Chimney-well design helps reduce cross-contamination
  • Designed to minimize air bubble formation
C-bottom plates feature a flat bottom for optical measurement, but the hard corners are curved to eliminate liquid traps and improve plate washing
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
MICROLON™ 200 and MICROLON™ 600 formats are available for consistent performance in binding assays. The chimney-well design option reduces the risk of cross-contamination and facilitates sealing by cap mat, thermal sealer, or adhesive sealer. Plates are supplied without lids.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty