Would you like to log in again?
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
655076 655077 655095 655074 655075 655097 655096 655094
82050-730CS 109.39 USD InStock
82050-730 82050-752 82050-754 82050-756 82050-724 82050-726 82050-728 82050-750
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
Microplates HTS Microplates
Black and white microplates are manufactured from virgin polystyrene resin and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Solid bottoms or clear bottoms
All of the black and white plates have square chimney wells with flat bottoms
LUMITRAC™ and FLUOTRAC™ formats are offered to allow a wide variety of applications, including cell-based assays. White LUMITRAC™ plates are for luminescence assays and black FLUOTRAC™ plates are for fluorescence assays.
Plates are available with solid or clear bottoms. Clear bottom plates are manufactured with µClear™ micro-thin polystyrene film, and are suggested for assays designed for luminescence and fluorescence detection in which imaging from the bottom is required. Plates are supplied without lids.
These 96-well polystyrene microplates plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty