Would you like to log in again?
CELLSTAR® Tissue Culture Plates, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CELLSTAR®
784080 655185 783092 781092 651160 655163 651161 781080-20 782092 650161 655162 650162 781079 781090-99 655083 655090-99 651182 675090 782078 655182 781086 781098 651162 781090 784086 675083 655073 650180 782073 651180 650185 675180 675086 782086 781091 781080 655161 782180 655086 655088 655180 655087 655160 655090 782093 781165 781182 781093 782080 781073 783160 655098 650160 655079
82051-276CS 189.64 USD InStock
82051-276 89085-314 82051-596 82051-398 82051-396 89135-050 82051-394 82051-392 82050-628 82051-438 82051-278 82050-633 82050-634 82050-758 89096-044 82051-390 82050-076 82051-288 82051-286 82051-282 89135-048 82050-048 82050-763 82050-764 82050-766 82050-722 82050-768 82051-280 82051-376 82050-050 82051-458 89047-272 82050-655 82050-732 82050-656 82050-658 82050-736 82050-052 82050-771 82050-772 75799-866 82051-266 82051-460 82051-304 82051-302 82051-424 82050-742 82050-744 82050-746 82050-625 82050-626 82050-748 82050-064 82050-661
CELLSTAR® Tissue Culture Plates, Greiner Bio-One
Microplates Cell Culture Plates
CELLSTAR® format sterile cell culture microplates are available tissue culture treated for the culture of anchorage-dependent cells, or untreated for suspension culture applications.
  • Designed to minimize wicking and reduce air bubble formation
  • Clear polystyrene film bottom
  • Chimney well design reduces cross-contamination
  • Certain models feature smaller working volumes to reduce sample usage
Multiwell culture plates come with a variety of well shapes
The 384-well plates with a maximum well volume of 30µL are special Small Volume™ plates that feature far smaller working volumes than typical 384-well plates, reducing sample usage while avoiding the instrument compatibility issues introduced by 1536-well plates. The wells on Small Volume™ plates feature conical well geometry; wells have a round top and a flat bottom, resembling a truncated cone.
Plates with µClear® bottoms feature a polystyrene microthin film bottom for clear imaging or viewing and minimized autofluorescence. HiBase plates are suitable for use with top-reading instruments, while LoBase plates are suitable for use with bottom-reading instruments. Dimensions: 128 L x 86 W mm (5 x 3⅜").
All plates are RNase- and DNase-free, noncytotoxic (EN ISO 10993), and nonpyrogenic. All plates have a Sterility Assurance Level (SAL) of 10-3 and have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty