Would you like to log in again?
CELLCOAT® Cell Culture Microplates, Greiner Bio-One
Supplier: Greiner Bio-One
CELLCOAT®
665940-005 655946 657940-005 781930 655950 657940 781936 655930 657930 781946 655948 662950-005 781956 662940 781948 655945 655944 655956 781947 781944 662950 662930 655936 655940 781940 665950-005 781945 662940-005 781950 657950 655955 657950-005
75796-002CS 93.33 USD InStock
75796-002 89218-660 75796-000 82051-352 82050-848 89047-254 82050-804 82050-904 89131-870 82050-806 89047-252 82050-906 82051-358 82050-908 75795-998 89047-270 82051-355 82051-356 82050-852 82050-798 82050-810 82050-812 75795-982 82050-850 82051-363 82051-360 89047-268 89047-266 82050-800 82050-802 75795-992 75796-008
CELLCOAT® Cell Culture Microplates, Greiner Bio-One
Microplates Cell Culture Plates
Convenient, ready-to-use polystyrene cell culture plates are coated with extracellular matrix (ECM) proteins or synthetic polypeptides.
  • Polystyrene construction
  • Compact footprint
  • Supplied with lid
  • Convenient and ready to use
These molecules provide an inert scaffolding that participates directly and indirectly in the regulation of cell growth and proliferation. Ideal for serum-free and serum-reduced cultivation or for differentiation of individual cell types. These multiwell tissue culture plates are consistently coated with Collagen Type 1 (from Rat Tail Tendon), Poly-D-Lysine or Poly-L-Lysine (both synthetic), and feature a “Best Before” date and lot number. All plates are supplied with lids and have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Dimensions: 128L x 86W mm
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty