Would you like to log in again?
96-Well Plate Lids, Greiner bio-one
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
656191 656178 656101 656172 656176 656161 656199 656190 691101 656170 656171 656197 691161
82051-582CS 216.85 USD InStock
82051-582 82050-838 82050-827 89131-671 82050-829 82050-840 82050-830 82050-832 82050-834 82050-823 82050-825 82050-836 82050-068
96-Well Plate Lids, Greiner bio-one
Lids Microplate Lids
Made of polycarbonate or polystyrene resin, these lids are best suited for chimney-well style 96-well plates in cell culture applications.
  • Polycarbonate or polystyrene resin construction
  • Fully skirted lids
  • Free of RNase and DNase
  • Nonpyrogenic
Half-area plates maintain ANSI/SBS standards and well-to-well pitch, yet feature a well diameter design that offers one-half the well surface area of standard plates
Standard full skirt lids allow easy gripping of the lid manually or robotically. Low-profile lids feature a partial skirt, facilitating robotic gripping of the plate while the lid remains on. Two corner notches on each lid assure proper plate-to-lid orientation, minimizing cross-contamination. Lids with condensation rings minimize both evaporation and cross-contamination. Black opaque lids are ideal when a loose cover is required to shield light-sensitive assays or compounds.
Lids are RNase-free, DNase-free, and nonpyrogenic.
This format allows a 50% sample volume reduction without moving to high well density plates. Half-area plates feature a medium binding polystyrene surface, which is ideal for work with nonpolar proteins and peptides.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty