Would you like to log in again?
CELLSTAR® Filter Cap Cell Culture Flasks, Greiner bio-one
Supplier: Greiner bio-one
660175 690175 658175 661175
82051-074CS 254.01 USD InStock
82051-074 82050-856 82050-878 82050-872
CELLSTAR® Filter Cap Cell Culture Flasks, Greiner bio-one
Flasks Cell Culture Flasks
A specific capillary pore membrane is used for the filter screw caps of CELLSTAR® filter cap cell culture flasks.
  • High airflow rate and optimal gas exchange
  • Additional standard screw caps, without filter, available on request
  • Improved cell adhesion through physical surface treatment
  • Cell culture flasks with filter screw caps
  • Canted neck
  • Graduation on both sides
  • Sterile and user-friendly packaging
  • 25cm², 75cm² and 175cm² cell culture flasks for variability of media volume
The defined pore diameter of 0.2 µm provides a sterile barrier against contamination
Due to the high airflow rate of the filter material, an optimal gas exchange is ensured.
Flasks are certified RNase-, DNase-free, and free of endotoxin (< 0.06EU/mL) for the most sensitive cell-based applications. Design of all flask formats allows efficient access to the cell lawn with cell scrapers and optimal access with pipettes. Flask options include 25, 75, and 182cm with plug-seal or filter (vented) caps. Filter cap flasks feature a 0.2 micron hydrophobic membrane for complete sterile gas exchange.
Flasks have raised rims for stacking. The 550mL flask has a low profile design. The 650mL flask has a high profile design. The red filter screw cap has a capillary 0.2µm pore diameter membrane and hydrophobic inner surface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty