Would you like to log in again?
SensoPlate™ 24-Well Glass-Bottom Plate, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SensoPlate™
662892
82050-898CS 289.18 USD InStock
82050-898
SensoPlate™ 24-Well Glass-Bottom Plate, Greiner Bio-One
Microplates HTS Microplates
Microplate exhibits a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
  • Ideal for applications requiring low autofluorescence
  • Exceptional optical clarity
  • Sterile
  • Chimney-well prevents cross-contamination
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plate consists of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame and features a chimney-well to help prevent cross-contamination. Plate is sterile, complies with SBS recommendations, and includes a lid.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty