Would you like to log in again?
CELLCOAT® Protein-Coated Petri Dishes, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
CELLCOAT®
628940 664950 628950 664940 628930 664910
82050-554CS 715 USD InStock
82050-554 82050-556 82050-920 82050-558 82050-922 82050-924
CELLCOAT® Protein-Coated Petri Dishes, Sterile, Greiner Bio-One
Culture Dishes Cell Culture Dishes
These high-quality, ready-to-use tissue culture dishes provide consistent and reproducible coatings with extracellular matrix (ECM) proteins or synthetic polypeptides.
  • Ensured consistent and reproducible coatings
  • Free of RNase and DNase
  • Nonpyrogenic
  • Provided sterile
These coatings provide scaffolding that participates directly and indirectly in the regulation of cell growth and differentiation. Dishes also offer a reliable alternative for applications involving sensitive cells. They are RNase-free, DNase-free, nonpyrogenic, and feature a "best before" date and lot number on each pack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty