Would you like to log in again?
CELLSTAR® Cell Culture Flasks with Standard Screw Cap, Greiner bio-one
Supplier: Greiner bio-one
658170 690170 661160 690160 660160
82050-854CS 161.82 USD InStock
82050-854 82050-876 82051-072 82051-070 82050-870
CELLSTAR® Cell Culture Flasks with Standard Screw Cap, Greiner bio-one
Flasks Cell Culture Flasks
Cell culture flasks with standard screw cap/plug seal (without filter).
  • Improved cell adhesion through physical surface treatment
  • Canted neck
  • Graduation on both sides
  • Sterile and user-friendly packaging
  • 25 cm², 75 cm², and 175 cm² growth areas
  • High and flat design of the 175 cm² cell culture flask for variability of media volume
All Greiner Bio-One cell culture flasks are made of high-grade polystyrene
Flasks are free of detectable DNase, RNase, and human DNA and are non-pyrogenic.
The 550 mL flask has a low profile design. The 650 mL flask has a high profile design.
For adherent cell culture, the surfaces of the standard cell culture flasks are physically surface-treated to improve cell adhesion and proliferation. The special design of the cell culture flasks makes it possible to efficiently reach the cell lawn with a cell scraper, whilst still providing optimal access with a pipette. The special neck design reduces the risk of wetting the inside of the cap with medium, thus providing additional protection against contamination. The stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator. Both sides have a printed graduation for easier filling.
Ordering information: Lot number is printed on each flask. Visual sterilization indicator on each box. Certificate of Quality in each case. For more information, including standard screw cap features (ventilation position option, etc.), please see the attached datasheets under the Related Information tab.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty