Would you like to log in again?
CELLSTAR© Suspension Culture Flasks, Screw Cap, Hydrophobic Surface, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
CELLSTAR®
658195 660190 690195 690190 658190 661195 661190
82051-078CS 243.11 USD InStock
82051-078 82050-874 82051-076 82050-858 82050-880 82050-860 82050-882
CELLSTAR© Suspension Culture Flasks, Screw Cap, Hydrophobic Surface, Sterile, Greiner Bio-One
Flasks Cell Culture Flasks
These graduated, canted-neck flasks are manufactured in an aseptic, fully automated facility from high-quality virgin polystyrene resin to ensure superior optical clarity.
  • High-quality virgin polystyrene construction
  • Superior optical clarity
  • Hydrophobic surface
  • Ideal for suspension cultures and hybridoma
Flask design makes it possible to efficiently reach the cell lawn with a cell scraper, while still providing optimal access with a pipet
All flasks are integrity tested to provide uniform growth and proliferation, and are sterilized by radiation. Flasks are nonpyrogenic, noncytotoxic, and free from DNase, RNase, and human DNA.
Flasks have a hydrophobic surface, ideal for suspension cultures and hybridoma. The 550 mL flask has a low profile design. The 650 mL flasks have a high profile design. Red filter screw cap has a capillary 0.2 µm pore diameter membrane and hydrophobic inner surface. Standard screw cap has a ventilation position.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty