Would you like to log in again?
Streptavidin-Coated Plates, Greiner Bio-One
Supplier: Greiner bio-one
781990 781996 781997 781995
82051-364CS 4026.16 USD InStock
82051-364 82051-370 82051-368 82051-366
Streptavidin-Coated Plates, Greiner Bio-One
Microplates Cell Culture Plates
Ideal for molecular biology applications, these plates feature low coating variance, high biotin binding capacity, and low streptavidin leaching.
  • Polystyrene construction
  • Available in a variety of clear or colored bottoms
  • Chimney-well design helps prevent cross-contamination
The very tight bond between streptavidin and biotin can be utilized to attach biotin-labeled proteins, nucleic acids, or other compounds to the microplate. Plates include a “Best Before” date and lot number on each pack. Available with black, white, clear, or µClear® bottom. The µClear® plates feature a reduced background, increasing sensitivity to 340nm. The 96-well plates feature a chimney-well design to prevent cross-contamination. 781990-589 and 781997-589 have blocker #5.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty