Would you like to log in again?
MASTERBLOCK® 96-Well Deep Well Microplates, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MASTERBLOCK®
780276-FD 780286-FD 780210 780261 780271-FD 780285-FD 780270-FD 780280-FD 780275-FD 786261 786201 780215 780201
82051-254CS 322.42 USD InStock
82051-254 82051-474 82051-244 82051-472 82051-242 82051-260 82051-238 89047-264 82051-228 82051-258 82051-248 82051-256 82051-246
MASTERBLOCK® 96-Well Deep Well Microplates, Greiner Bio-One
Microplates Deep Well Microplates
Microplates are manufactured from virgin polypropylene resin, and feature uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
  • Virgin polypropylene construction
  • Alphanumeric coding
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Autoclavable
Microplates are biologically inert, exhibit low binding characteristics, offer a high temperature tolerance (–196 to 121°C/–321 to 250°F), and are resistant to many standard laboratory chemicals
Microplates feature alphanumerically coded wells. Conical well tops aid in complete sample recovery. Plates are autoclavable and supplied without lids.
Microplates are RNase- and DNase-free, and nonpyrogenic. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
These properties provide optimal conditions for laboratory specimen and compound storage. Ideal for cultivating bacteria or yeasts.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty