Would you like to log in again?
384-Well Small Volume™ Assay Plates, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Small Volume™
784075-25 788095 784076 784101 788101 784076-25 788096 784077 784074 784075
89085-324CS 543.21 USD InStock
89085-324 82051-486 82051-442 82051-450 82051-484 82051-462 89085-330 82051-456 89085-328 82051-444
384-Well Small Volume™ Assay Plates, Greiner Bio-One
Microplates HTS Microplates
Microplates provide high-throughput assay miniaturization in a 384-well system, thereby avoiding the instrument compatibility issues introduced by 1536-well plates while still reducing sample usage.
  • Suited for high-throughput assays
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Supplied sterile
  • Conical well geometry
All plates are manufactured from virgin polystyrene resin, except for the black UV-Star® 384-Well Microplates, which is constructed of cycloolefin copolymer (COC) for better chemical resistance and detections in lower UV wavelengths
LUMITRAC™ treatment is for luminescence assays, and FLUOTRAC™ treatment is for fluorescence assays. HiBase plates are suitable for all standard high throughput applications. LoBase plates feature a chimney-like arrangement of wells in which each well stands on its own to reduce the possibility of cross-contamination. Chimney wells also feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer. LoBase plates are available with µClear® polystyrene microthin film bottom for clearer imaging or viewing and minimized autofluorescence. Plates are supplied without lids. The standard surface treated LoBase plates are supplied sterile. The black standard LoBase Plates is supplied with lid.
Plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Wells feature conical geometry, in which wells have a round top and a flat bottom, resembling a truncated cone. Plates are compatible with microplate handling equipment and robotics.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty