Would you like to log in again?
SensoPlate™ 1536-Well Glass-Bottom Plates, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SensoPlate™
783892P 783892
82051-562CS 56.62 USD InStock
82051-562 82051-440
SensoPlate™ 1536-Well Glass-Bottom Plates, Greiner Bio-One
Microplates HTS Microplates
Microplates exhibit a bottom flatness better than 100 µm to offer high-quality performance for all applications where low autofluorescence and exceptional optical clarity are required.
  • Ideal for applications requiring low autofluorescence
  • Exceptional optical clarity
  • Sterile
  • Supplied with a lid
Ideal for high-resolution imaging, sensitive fluorescence, and confocal microscopy such as single molecule detection (SMD). Transmission measurements are capable in the wavelength range above 350nm. Plates consist of a 175 µm thick borosilicate glass-bottom plate bonded with a black polystyrene frame. Plates are sterile, comply with SBS recommendations, and include a lid.
Ordering information: For 1536-well SensoPlate™ Plus glass-bottom plates, see 82051-620 series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty