Would you like to log in again?
96-Well Half Area Microplates, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
675161 675074 675001 675077 675075 675101 675061 675076 675096
82051-574CS 211.36 USD InStock
82051-574 82050-060 82050-062 82050-040 82050-042 82050-044 82050-056 82050-046 82051-576
96-Well Half Area Microplates, Greiner Bio-One
Microplates HTS Microplates
Polystyrene half area microplates are compatible with manual and automated pipetting, and dispensing and reading devices used with standard 96-well plates.
  • Polystyrene construction
  • Chimney-well design helps reduce cross-contamination
  • Clear film bottoms
  • Suitable for cell-based assays
This design allows up to a 50% sample and reagent reduction and the flat bottoms allow stackability with or without lids
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardized path-length, eliminating the need for path-length determinations. MICROLON™ formats are ideal for consistent performance in binding assays. White LUMITRAC™ plates are ideal for luminescence assays, while black FLUOTRAC™ plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
The chimney style well design reduces the risk of cross-contamination and aids sealing by cap mat, thermal sealer, or adhesive sealer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty