Would you like to log in again?
HELL GROUND WOMEN
Supplier: DESCO
Foot Grounder Toe Women^ Foot Grounder, Toe, Women 30 in Light Blue Tab, 2 Meg Resistor, Features: Toe cup, Non-marring interior, D-ring fastening system, 2-ply high abrasion, wear resistant rubber, 8 strand nylon PE grounding tab, Exceptionally tear resi
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HELL GROUND WOMEN 82128-786 07595 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty