Would you like to log in again?
White SMS Coveralls with Attached Hood, Elastic Wrist and Ankle, International Enviroguard™
Supplier: International Enviroguard
2225-L 2225-XL 2225-M 2225-4XL 2225-3XL 2225-2XL
83007-080CS 196.61 USD InStock
83007-080 83007-078 83007-074 83007-076 83007-082 83007-072
White SMS Coveralls with Attached Hood, Elastic Wrist and Ankle, International Enviroguard™
Disposable Garments
SMS Coverall provide an excellent particulate barrier along with protection from light liquid splashes and sprays.
  • SMS fabric is strong yet breathable to keep users cool and help avoid heat stress
  • Storm flap over zipper adds an extra layer of protection
  • Attached hood with elastic wrists and ankles for snug, secure fit
Three-layer SMS is lightweight and breathable. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with user for more comfort and better tear resistance.
Recommended applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations, and more.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty