Would you like to log in again?
Threadlocker 271™ High Strength Adhesive, Loctite®, Henkel
Supplier: Henkel
Threadlocker 271™
27131
83007-504CS 628 USD InStock
83007-504
Threadlocker 271™ High Strength Adhesive, Loctite®, Henkel
Adhesives
Designed for the permanent locking and sealing of fasteners such as large bolts and studs (M25 and larger) up to 25mm (1") diameter.
  • Acrylic threadlocker
  • High-strength
  • Low-viscosity
The adhesive cures when confined in the absence of air between close fitting metal surfaces and prevents loosening and leakage from shock and vibration. Low viscosity reduces the migration of liquid adhesive after application.
Certifications: Adhesive meets MIL-SPEC (S-46163A) for existing designs and ASTM D5363 for new designs. CFIA certified. UL listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty