Would you like to log in again?
ceramus™-classic Bottle-Top Dispensers, Hirschmann
Supplier: Hirschmann Laborgeräte
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ceramus™
9351100 933110010 931000010 932000010 9361100 933000010 934110010 934000010 932110010 931110010 9360000 9350000
83007-610EA 906.06 USD InStock
83007-610 83007-632 83007-622 83007-612 83007-624 83007-630 83007-620 83007-618 83007-614 83007-626 83007-616 83007-628
ceramus™-classic Bottle-Top Dispensers, Hirschmann
Bottle Top Dispensers
Dispensers feature a ceramic plunger that is made of solid, 99.7% pure aluminum oxide, which guarantees the highest degree of chemical resistance, without abrasion or swelling
The cylinder is made of calibrated DURAN® (borosilicate 3.3) glass, is predominantly inert and wear-free, and is protected by robust plastic sheathing. The protective sheathing incorporates two glass windows so that air bubbles can be easily identified. The check valves are also predominantly inert and wear-free. The ball and seat are made of borosilicate glass, the valve springs are made of Hastelloy® material, the valve block and housing are made of ECTFE copolymer, and the suction and dispensing tubes are made of FEP (fluorinated ethylene propylene). All components can be disassembled quickly and easily for cleaning, and can be autoclaved at 121°C (250°F).
The suction and dispensing valves can be easily exchanged through the use of an assembly key. The suction tube is screw-threaded for an airtight seal. The dispensing tube is housed within a stable support tube to ensure splash-free dispensing. Every component that comes in contact with the medium is uncolored, free of additives, and manufactured from a material that is inert against the most common media.
These dispensers are equipped with a digital volume setting system, an internal step gauge, and a click-stop micro-step volume adjustment dial. The dial guarantees reproducible results and is calibrated for the lifetime of the device. Dispensers are not for use with hydrofluoric acid.
Cap size: 32 mm
Ordering information: Manufacturer’s one-year warranty. Each unit includes a suction tube, three thread adapters (A28, A45, and S40), and an individual quality control certificate that documents the serial number and five test values, and confirms the conformity according to weights and measures regulations. Units that dispense volumes of up to 10mL are supplied with a 946mL (32oz.) amber glass bottle.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty