Would you like to log in again?
ceramus™ Bottle-Top Dispensers, Hirschmann
Supplier: Hirschmann Laborgeräte
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ceramus™
9342300 934210010 9362000 9352000 934200010 933210010 932210010 933200010 931200010 9362100 932200010 9352100 931210010
83007-654EA 1349.14 USD InStock
83007-654 83007-644 83007-634 83007-656 83007-646 83007-650 83007-640 83007-652 83007-642 83007-658 83007-636 83007-648 83007-638
ceramus™ Bottle-Top Dispensers, Hirschmann
Bottle Top Dispensers
Dispensers feature a ceramic plunger that is made of solid, 99.7% pure aluminum oxide, which guarantees the highest degree of chemical resistance, without abrasion or swelling.
Dispensers feature an internal step gauge and a click-stop micro-step volume adjustment dial that guarantees reproducible results, and is calibrated for the lifetime of the device
The cylinder is made of calibrated DURAN® (borosilicate 3.3) glass and is, like the valves, predominantly inert and wear-free. The ball and seat are made of 3.3 borosilicate glass, the valve springs are made of Hastelloy® material, the valve block and housing are made of ECTFE (ethylene chlorotrifluoroethylene) copolymer, and the suction and dispensing tubes are made of FEP (fluorinated ethylene propylene). The robust suction tube is tightly screwed to the device, which prevents air bubbles from forming during suction.
The ceramus HF unit (83007-658) is specifically designed for dispensing hydrofluoric acid and other problematic media. This unit features ceramic components instead of glass.
A reagent recirculation system ensures quick, simple priming and prevents reagent waste. Units feature a Luer-lock connection for a filter or dry tube; a strong, stable discharge tube with cap; a protective housing; a plunger handle with ergonomic grip; and 360° rotation so that labels or lettering on the bottle can be turned to always face the user. The complete device can be autoclaved at 121°C (250°F). All parts can be easily disassembled.
Ordering information: Each unit includes a suction tube, recirculation tube, wrench to connect/disconnect the valves, three thread adapters (A32, A38, and S40), and an individual quality control certificate that documents the serial number and five test values, and confirms the conformity according to weights and measures regulations. Dispensers that accommodate volumes of up to 10mL are supplied with a 946mL (32oz.) amber glass bottle. Dispensers capable of using hydrochloric acid with trace metals analysis are available; contact your VWR sales representative for more information.
Caution: Note: Dispensers are not intended for use for trace metals analysis with hydrochloric acid (with the exception of dispenser 83007-658).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty