Would you like to log in again?
OPTI-LYNX™ In-Line Trap / Filter Holder Kit
Supplier: Optimize Technologies
OPTI-LYNX™
11-02225-AA
83008-166EA 855.68 USD InStock
83008-166
OPTI-LYNX™ In-Line Trap / Filter Holder Kit
Chromatography Supplies Chromatography Spare Parts
The OPTI-LYNX™ In-Line Holder can be connected to ¹/₁₆" tubing for in-line filtration or trapping anywhere on your HPLC system.
  • For use with OPTI-LYNX™ Traps and Filters
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection
  • Hardware is only plumbed once
  • Hand-tight - no tools required
The OPTI-LYNX™ trapping configuration can also be placed directly within the loop of an injection valve to facilitate automated loading, flushing and elution. With Opti-Lynx, accessing and changing the insert takes only a quarter turn. Replace traps, filters or change configurations in seconds - without tools. OPTI-LYNX™ traps and filters (ordered separately) come in several bed volume/ diameter/ porosity combinations, allowing you to find your own balance between capacity and volume.
Delivery information: Includes bayonet connector, holder cap, holder tube and fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty