Would you like to log in again?
OPTI-LYNX™ Direct-Connect Trap/Pre-Column Filter Holder Kit
Supplier: Optimize Technologies
OPTI-LYNX™
11-02226-AA
83008-168EA 784.29 USD InStock
83008-168
OPTI-LYNX™ Direct-Connect Trap/Pre-Column Filter Holder Kit
Chromatography Supplies Chromatography Spare Parts
The OPTI-LYNX™ Direct-Connect Trap/Pre-Column Holder System comes complete with the hardware you need to quick-connect to any 10 to 32 valve/port for trapping or analytical column for pre-column filtration.
  • For use with OPTI-LYNX™ Traps and Filters
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection to any column brand or 10 to 32 injection valve
  • Hardware is only plumbed once
  • Hand-tight - no tools required
With Opti-Lynx, accessing and changing the insert takes only a quarter turn. Replace traps, filters or change configurations in seconds, without tools. OPTI-LYNX™ traps and filters (ordered separately) come in several bed volume/ diameter/ porosity combinations, allowing you to find your own balance between capacity and volume.
Delivery information: Includes: Universal port adapter, holder cap, holder tube and fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty