Would you like to log in again?
Reagent Bottles with Stoppers, Polypropylene, Narrow Mouth, BrandTech
Supplier: BrandTech
V100594 V100394 V100494
83008-520PK 150.16 USD InStock
83008-520 83008-512 83008-516
Reagent Bottles with Stoppers, Polypropylene, Narrow Mouth, BrandTech
Bottles Plastic Bottles
Polypropylene reagent bottles with polypropylene stoppers provide excellent chemical resistance to many aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
  • Made of durable polypropylene
  • Tight fitting stoppers
  • Conical shoulders
Bottle 83008-532 has a handle. Bottles and stoppers are autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty