Would you like to log in again?
VITLAB® Reagent Bottles, PP with NS Stoppers, BrandTech
Supplier: BrandTech
V100694
83008-524PK 216.16 USD InStock
83008-524
VITLAB® Reagent Bottles, PP with NS Stoppers, BrandTech
Bottles Plastic Bottles
Slope-shouldered polypropylene bottles feature NS tapered polypropylene stoppers to help prevent leakage.
  • Break-resistant
  • Polypropylene bottle and stopper for high chemical resistance
  • High translucency
  • Rugged design
5 liter size includes a handle to facilitate movement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty