Would you like to log in again?
VITLAB® Reagent Bottles, with PP Screw Caps, BrandTech
Supplier: BrandTech
V100689 V100889 V100989 V100789 V100489 V100389
83008-530PK 66.79 USD InStock
83008-530 83008-534 83008-510 83008-522 83008-514 83008-526
VITLAB® Reagent Bottles, with PP Screw Caps, BrandTech
Bottles Plastic Bottles
Slope-shouldered polypropylene bottles feature polypropylene screw caps.
  • Bottles and caps are autoclavable
They provide excellent chemical resistance to aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
Ordering information: For replacement caps, see 83009-032 series.
Delivery information: 10 liter size includes two handles, five liter size includes a handle to facilitate movement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty