Would you like to log in again?
VITLAB® Volumetric Flasks with Screw Caps, PFA, BrandTech
Supplier: BrandTech
V107597 107497 107297 107397 107097 V107197
83008-574PK 1083.49 USD InStock
83008-574 83008-573 83008-566 89093-094 83008-569 83008-571
VITLAB® Volumetric Flasks with Screw Caps, PFA, BrandTech
Flasks Volumetric Flasks
Flasks feature a durable, easy-to-read, individually calibrated ring marking.
  • PFA for extreme chemical resistance
  • Screw caps help maintain sample integrity
Highly translucent, hydrophobic surfaces ensure easy meniscus verification. Made of PFA (perfluoroalkoxy copolymer). Highly heat- and chemical-resistant, autoclavable, and easy-to-clean. Can withstand temperatures from –200 to 260 °C (–328 to 500 °F). PFA screw caps help protect against outside contamination.
Certifications: Flasks meet Class A tolerances per DIN EN ISO 1042.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty