Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sample Containers with Screw Caps, Polypropylene, BrandTech

Supplier: BrandTech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


V130394 V130594
83008-660PK 251.38 USD InStock
83008-660 83008-668
Sample Containers with Screw Caps, Polypropylene, BrandTech
Sample Containers General Purpose Containers
Translucent polypropylene containers with screw caps

  • Chemically resistant polypropylene
  • Easy to clean
  • Screw on caps for security

Maximum operating temperature: 125 °C (225 °F). Autoclavable.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty