Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VITLAB® Griffin Beakers, PMP, BrandTech
Supplier: BrandTech
V61803 V61603 V60803 V61503 V60903 V61103 V60703 V61203 V60503 V61703 V61403 V61003 V60603
83008-808PK 127.85 USD InStock
83008-808 83008-780 83008-784 83008-792 83008-820 83008-832 83008-788 83008-796 83008-824 83008-800 83008-828 83008-804 83008-816
VITLAB® Griffin Beakers, PMP, BrandTech
Beakers
High-quality beakers made of plastic polymethylpentene with moderately good chemical resistance.
  • Glass-clear polymethyl pentene (PMP)
  • Red screened graduations for high contrast with many liquids
Beakers feature molded graduations with red-screened overprint for legibility.
Certifications: All beakers meet the requirements of ISO 7056.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty