Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VITLAB® Griffin Beakers, Polypropylene, BrandTech
Supplier: BrandTech
V607081 V609081 V608081 V605081 V617081 V616081 V615081 V614081 V606081 V612081 V610081 V611081 V618081
83008-790PK 58.3 USD InStock
83008-790 83008-794 83008-782 83008-798 83008-810 83008-830 83008-786 83008-802 83008-822 83008-834 83008-806 83008-818 83008-826
VITLAB® Griffin Beakers, Polypropylene, BrandTech
Beakers
High quality beakers feature molded graduations (except 83008-814) with blue-screened overprint for legibility.
  • Highly chemically resistant polypropylene
  • High translucency
  • Blue screened graduations provide high contrast with many liquids
Beakers are translucent for compatibility and offer good chemical and break resistance.
Certifications: All beakers meet the requirements of ISO 7056.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty