Would you like to log in again?
VITLAB® Graduated Cylinders, Polypropylene, Class B, BrandTech
Supplier: BrandTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V652941 V649941 V653941 V648941 V646941 V651941 V647941 V650941
83008-894PK 181.4 USD InStock
83008-894 83008-864 83008-884 83008-874 83008-934 83008-924 83008-914 83008-904
VITLAB® Graduated Cylinders, Polypropylene, Class B, BrandTech
Cylinders Measuring Cylinders
Made of polypropylene with ring-molded graduations at major increments to simplify measurements.
  • Break-resistant
  • High translucency
  • Polypropylene for high chemical resistance
  • Hexagonal base resists rolling
Class B per DIN 12681.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty