Would you like to log in again?
VITLAB® Graduated Cylinders, Polypropylene, Class B, Blue Scale, BrandTech
Supplier: BrandTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V648081 V647081 V649081 V652081 V653081 V651081 V650081 V646081
83008-860PK 133.31 USD InStock
83008-860 83008-880 83008-870 83008-930 83008-920 83008-910 83008-900 83008-890
VITLAB® Graduated Cylinders, Polypropylene, Class B, Blue Scale, BrandTech
Cylinders Measuring Cylinders
Made of polypropylene with ring-molded graduations overprinted with blue screening to simplify measurements.
  • Translucent polypropylene is chemically resistant
  • Blue screening for high contrast with many liquids
These VITLAB® graduated cylinders are made of polypropylene for durability and chemical resistance. Cylinders are highly translucent with molded graduations, and blue-screened overprint for easy meniscus readings. Hexagonal base resists rolling.
Certifications: Class B per DIN 12681.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty