Would you like to log in again?
VITLAB® Volumetric Flasks with [ST] Stoppers, Class A and Class B, BrandTech
Supplier: BrandTech
67104 V67495 67204 67604 673941 V67295 67304 V67195 675941 V67695 671941 672941 67404 V67595 V67395 677941 674941 676941 67704 V67604 67504
89093-097PK 233.6 USD InStock
89093-097 83009-004 83009-005 83009-009 76552-220 83008-996 83008-985 83008-975 83008-986 83008-995 83008-989 83008-979 83008-976 83008-965 83008-966 83008-999 89093-105 76552-212 83008-969 83008-959 76552-214
VITLAB® Volumetric Flasks with [ST] Stoppers, Class A and Class B, BrandTech
Flasks Volumetric Flasks
Individually calibrated to contain at 20 °C (68 °F).
  • Volumetric flasks with traditional push in stoppers
  • Class A (PMP) or Class B (PMP or PP)
  • Class A flasks are supplied with a batch certificate
Flasks feature durable ring graduation marks. Made of break-resistant, translucent, autoclavable, easy-to-clean PMP (polymethylpentene) or polypropylene. Supplied with a [ST] stopper.
Certifications: Flasks meet either Class A or Class B tolerances per DIN EN ISO 1042.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty