Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VITLAB® Forceps, PMP, BrandTech

Supplier: BrandTech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


V67895 V67995
83009-010PK 64.36 USD InStock
83009-010 83009-011
VITLAB® Forceps, PMP, BrandTech
Forceps
Polymethylpentene (PMP) forceps with pointed ends.

  • Autoclavable at 121 °C for 20 minutes
  • Translucent

PMP is suitable for autoclaving and microwaves. Resistant to aliphatic alcohols, aldehydes, ester, aliphatic hydrocarbons, ketones, alkalis, and most acids (except oxidizing acids). Not recommended for use with ether, aromatic hydrocarbons, halogenated hydrocarbons, and oxidizing agents. Color: transparent white.

Operating temperature: 0 to 150°C (32 to 302°F)

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty