Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VITLAB® Forceps, POM, BrandTech

Supplier: BrandTech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


V68099 V68399 V68199 V68299
83009-012PK 28.68 USD InStock
83009-012 83009-013 83009-014 83009-015
VITLAB® Forceps, POM, BrandTech
Forceps
Polyoxymethylene (POM) forceps with rounded ends.

  • Rounded ends
  • Opaque orange-yellow

POM is suitable for autoclaving but not suitable for microwaves. Frequent autoclaving may reduce mechanical stability. Resistant to aliphatic alcohols, ether, aldehydes, aliphatic hydrocarbons, aromatic hydrocarbons, halogenated hydrocarbons, ketones, and alkalis.

Operating temperature: –40 to 130°C (–40 to 266°F)

Caution: Not recommended for use with ester, acids, and oxidizing agents.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty