Would you like to log in again?
VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Supplier: BrandTech
VITLAB®
V92189 V92089 V91789 V91989
83009-054PK 209.72 USD InStock
83009-054 83009-050 83009-052 83009-053
VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Bottles Plastic Bottles
General-use, high-density polyethylene laboratory bottles come with polypropylene screw caps.
  • Space-saving shape
  • HDPE construction with PP screw caps
  • Narrow mouth
Bottles provide excellent chemical resistance to aliphatic and aromatic hydrocarbons, alcohols, aldehydes, alkalis, and acids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty