Would you like to log in again?
VITLAB® Laboratory Bottles, LDPE, Narrow Mouth, with Screw Caps, PP, BrandTech
Supplier: BrandTech
V94989 V94689 V95089 V94589
83009-067PK 145.35 USD InStock
83009-067 83009-069 83009-071 83009-073
VITLAB® Laboratory Bottles, LDPE, Narrow Mouth, with Screw Caps, PP, BrandTech
Bottles Plastic Bottles
Round-shouldered general-use low-density polyethylene laboratory bottles come with polypropylene screw caps.
  • LDPE with PP screw caps
  • Round shoulder design
  • Narrow mouth
Bottles provide excellent chemical resistance to aliphatic alcohols, aldehydes, alkalis, and acids.
Ordering information: For replacement caps, see 83009-032 series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty