Would you like to log in again?
SOCOREX® ACURA® Manual 815 Single-Channel Pipettors With Tip Ejectors, Fixed Volume, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Socorex™ Acura®
W810060 W810034 W810022 W810062 W810042 W810010 W810052 W810016 W810018 W810012 W810020 W810036 W810054 W810056 W810024 W810004 W810008 W810000 W810038 W810032 W810026 W810002 W810028 W810014 W810048 W810040 W810030 W810044 W810046 W810050 W810006 W810058
83009-098EA 517.53 USD InStock
83009-098 83009-154 83009-110 83009-132 83009-134 83009-112 83009-114 83009-136 83009-116 83009-138 83009-118 83009-140 83009-120 83009-142 83009-144 83009-100 83009-122 83009-102 83009-146 83009-124 83009-126 83009-104 83009-148 83009-128 83009-106 83009-108 83009-092 83009-094 83009-150 83009-096 83009-152 83009-130
SOCOREX® ACURA® Manual 815 Single-Channel Pipettors With Tip Ejectors, Fixed Volume, DWK Life Sciences
Pipettes Micropipettes
Lightweight pipettors are designed for precise, accurate pipetting of solutions, reagents, acids, serums, and other liquids, and for gel-sequencing applications.
  • Dispensing is accomplished with a single plunger stroke
  • Stainless steel and PVDF plunger assembly is chemically resistant
  • Autoclavable to 121 °C (250 °F) without disassembly or recalibration
A differentiated overshoot function and separate push button for tip ejection reduce dead volume and prevent accidental de-tipping. The glove-friendly JustTip™ adjustable tip ejector allows the use of most pipette tips.
Ordering information: Supplied with a manufacturer’s three-year warranty, including a two-year warranty on the original calibration. The 2000 µl pipettor includes an adapter for use with a Pasteur pipet. Pipettors 83009-148, -150, -152, and -154 are supplied with polypropylene fiber filters that may be used with polypropylene tips to prevent cross-contamination.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty