Would you like to log in again?
Charging Stand for Socorex® ACURA® Electronic Pipettes
Supplier: Socorex
Socorex™ Acura®
W820019
83009-242EA 487.54 USD InStock
83009-242
Charging Stand for Socorex® ACURA® Electronic Pipettes
Pipette Stands
This stand allows charging and storing of up to three electronic pipettes or battery packs simultaneously.
  • Each position is fitted with a red/green charging LED
  • Charging stand supplied with every Acura® electro basic package
Replacement battery packs suitable for all Acura® electro pipettes.
Ordering information: Charging stand does not include the 100-240V power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty