Would you like to log in again?
Super Erecta®/Super Adjustable Super Erecta® Top-Track™ High-Density Storage Systems, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
TTE24C LTTE24C TTM21S TTM24C TTE18K3 TTM21S LTTA24C LTTE18C TTM18K3 TTE21S TTM21K3 TTA21C TTE24C TTE18S TTA18K3 TTE24S TTM18K3 TTM18S TTE21S TTM24K3 TTM24C TTA24K3 LTTE21C LTTE21C LTTM18C TTE24K3 LTTE24C LTTM21C TTA18C TTE21C TTE24S TTA24K3 TTA18S LTTM18C LTTA21C TTE21K3 TTE24K3 LTTM24C TTA18S TTE18S TTA18C TTE18C TTM18C LTTA24C LTTE18C TTA21K3 TTE18C TTM18C LTTM24C TTA21C TTA24C TTE21C LTTA21C LTTM21C TTE21K3 TTM18S LTTA18C TTA24S TTM21C TTA24S TTA18K3 TTA21S TTM24K3 TTE18K3 TTA21S TTA24C TTM21C LTTA18C TTM21K3 TTA21K3
83009-472EA 1368.4 USD InStock
83009-472 83009-550 CA83009-482 83009-474 CA83009-560 CA83009-484 83009-552 CA83009-562 83009-476 CA83009-486 83009-554 83009-478 CA83009-520 CA83009-564 CA83009-488 83009-556 CA83009-566 CA83009-522 83009-558 CA83009-568 CA83009-524 CA83009-526 83009-518 CA83009-528 83009-480 83009-482 83009-540 CA83009-472 CA83009-550 CA83009-474 83009-542 CA83009-552 CA83009-476 83009-544 CA83009-554 83009-546 CA83009-478 CA83009-556 83009-548 CA83009-558 CA83009-518 CA83009-480 83009-530 CA83009-540 83009-532 CA83009-542 83009-534 CA83009-544 83009-536 CA83009-546 83009-538 CA83009-548 83009-560 83009-484 83009-562 83009-486 83009-564 83009-520 83009-488 CA83009-530 83009-566 83009-522 CA83009-532 83009-524 83009-568 CA83009-534 83009-526 CA83009-536 83009-528 CA83009-538
Super Erecta®/Super Adjustable Super Erecta® Top-Track™ High-Density Storage Systems, Metro™
Furniture Shelves
Top-Track™ system increases storage capacity up to 50% and maximizes the usable storage area in a given space.
  • Chrome-plated units are ideal for dry environments with minimal exposure to corrosives
  • Type 304 stainless steel units provide maximum protection in corrosive environments
Metroseal 3™ units are made of electroplated carbon steel with a unique corrosion-resistant epoxy, an economical alternative to stainless steel units, and are coated with antimicrobial product protection which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Stationary end and stationary intermediate unit kits for single systems include four 218.4 cm (86") high posts and the necessary hardware for connecting track sets to two stationary end units or one stationary intermediate unit. Stationary end and stationary intermediate unit kits for double deep systems include eight 218.4 cm (86") high posts, the necessary hardware for connecting two stationary end units in the double deep configuration, and the necessary hardware for connecting track sets to two stationary end units or one stationary intermediate unit. A stationary intermediate unit is required when track length exceeds 6.4 m (21'). The stationary end and stationary intermediate units must contain a minimum of four shelves. The width of the stationary end and stationary intermediate units should be equal to or greater than the width of the mobile units.
Mobile unit kits for single systems include four 188 cm (74") posts, four casters with bumpers, four caster channels, and four roller assemblies. Mobile unit kits for double deep systems also include the necessary hardware to connect two mobile units in the double deep configuration. The recommended maximum load rating for a mobile unit is 408.2 kg (900 lbs.). For the mobile units to function properly, the floor area should be level, smooth, and free from large cracks and raised obstacles.
Track sets include two segments of track. Only one track set is required between stationary units. The wall mount bracket kit includes one pair of brackets to attach a Top-Track™ system to a wall. The stop plate kit includes stops and fasteners for one end of the track set.
Note: Super Erecta® or Super Erecta® Super Adjustable™ shelves must be purchased separately.
The overhead track system guides mobile units between stationary units. An access aisle can be opened between any two units as needed. This open access to all units promotes cleanliness and permits users to walk or push utility carts in between units, simplifying the material handling task.
Ordering information: Metroseal 3™ products feature manufacturer's twelve-year limited warranty against rust formation. For chrome-plated Super Erecta® shelves, see 19766-070 series. For stainless steel Super Erecta® shelves, see 19766-072 series. For Metroseal 3™ Super Erecta® shelves, see 83009-332 series. For Super Erecta® Super Adjustable™ shelves, see 22233-716 series. Additional accessories such as shelf dividers, stackable ledges, tray slides, label holders, and more are also available; contact your VWR representative for more information.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty