Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Whatman™ Mini-UniPrep Syringeless Filters
Supplier: Whatman products (Cytiva)
Whatman™ Mini-UniPrep™ Mini-UniPrep™
UN503NPUORG US203NPEAQU UN203NPUDPP UN203NPEPES US203NPENYL UN503NPEPP UN203APUAQU UN503NPEORG UN503NPUORG UN203APEORG UN203NPUAQU US203NPUAQU UN203APEPES UN203APEAQU US203APUNYL UN203NPEAQU UN503NPUPES US503NPEAQU UN203APEPP UN203APEPP UN203NPENYL US503NPUORG UN503NPUPP UN503NPUPP UN203APEPES US203APUNYL US203NPUNYL UN203NPEAQU UN203NPUPES US503NPEPES UN203NPUPP UN503NPURC UN203APUORG UN503NPUGMF UN503NPEORG US203NPEAQU UN503NPUNYL US203NPUPES UN503NPENYL US203NPUDPP UN203APENYL US503NPEPP UN203NPUPES UN203APEORG UN203NPERC US203NPUDPP UN203NPUGMF UN203NPENYL UN203NPURC US503NPUDPP UN203APUPP UN203APUDPP US203NPEORG UN503NPUAQU US503NPUGMF UN203NPERC UN203NPEORG US503NPEAQU UN503NPUGMF UN203NPUORG UN503NPUNYL US203NPUORG UN203NPEPP UN203NPUORG US503NPUORG US203NPENYL US503NPEPES US203NPUGMF UN503NPUAQU UN503NPEAQU US503NPEORG US203NPUGMF US203NPUPP UN203NPUGMF US503NPUAQU UN503NPERC UN203APUAQU UN203APUPP UN203APEAQU US503NPUPP UN203APUGMF UN203APUDPP UN203NPEORG US203NPEPES US203NPUPP UN203APUORG US203NPUPES US203NPEORG US503NPUAQU UN203NPEPES US503NPENYL US203NPEPES US203NPEPP US503NPEPP UN203NPUAQU UN203NPEPP US503NPUDPP UN203NPUNYL UN203NPURC UN203NPUNYL UN503NPEAQU US503NPEORG US203NPUAQU UN503NPUPES US203NPEPP UN203APUGMF UN203APUNYL UN203APENYL UN203APUNYL UN503NPUDPP UN503NPUDPP UN203NPUPP US203NPUORG US203NPUNYL US503NPUPP US503NPUGMF UN503NPURC UN203NPUDPP UN503NPENYL US503NPENYL UN503NPERC
28137-772PK 4405.65 USD InStock
28137-772 CA89233-788 84009-510 CA89233-786 28137-770 84009-512 84009-514 84009-516 28137-776 CA83009-802 CA83009-806 CA83009-808 CA12000-540 12000-530 CA12000-542 12000-532 12000-536 12000-538 CA14224-914 CA12000-530 12000-524 CA12000-532 12000-526 12000-528 CA12000-536 CA12000-538 28137-750 CA28137-760 28137-752 CA28137-762 14224-762 83009-810 28137-758 CA28137-768 CA83009-820 83009-812 CA83009-822 CA14224-778 28137-754 83009-814 CA83009-824 CA28137-764 28137-756 CA14224-930 83009-816 CA28137-766 CA83009-826 83009-818 14224-914 97034-680 CA12000-524 CA12000-526 CA10147-936 CA12000-528 CA28137-770 28137-762 CA28137-772 28137-764 28137-760 14224-930 CA83009-810 83009-802 CA83009-812 28137-766 CA83009-814 14224-778 CA28137-776 28137-768 83009-806 CA83009-816 83009-808 CA83009-818 83009-830 83009-798 CA14224-794 83009-834 14224-946 CA14224-946 CA28137-750 14224-794 83009-820 CA83009-830 CA28137-756 83009-822 CA83009-798 CA28137-758 83009-824 CA83009-834 CA28137-752 83009-826 CA28137-754 CA97034-680 97015-564 97015-562 10147-936 CA14224-730 CA84009-504 CA14224-976 CA84009-508 89233-790 CA84009-506 CA97015-564 CA97015-562 CA84009-510 89233-786 89233-788 97055-768 84009-504 CA84009-514 CA89233-790 CA84009-512 84009-506 14224-730 84009-508 14224-978 14224-976 CA84009-516 CA14224-978 12000-540 12000-542 CA97055-768
Whatman™ Mini-UniPrep Syringeless Filters
Filters Syringeless Filters
The Whatman Mini-UniPrep Syringeless Filters combine four products into one syringeless filter solution for efficient chromatography sample preparation for a broad range of applications. They are built for fast and easy high performance liquid chromatography/ultra high performance liquid chromatography sample preparation and analysis.
  • All-in-one filtration device for quick and cost-effective chromatography sample processing
  • Eliminates the need for separate syringes, syringe membrane filters, vials, and septa
  • Designed for use with an UHPLC or HPLC autosampler for high-throughput analysis
  • 0.2 and 0.45 µm filter pore sizes available to meet filtration sample requirements
  • Compatible with most major autosamplers
  • Housing options: amber to prevent photodegradation of light-sensitive samples, or translucent for easy visual inspection made from polypropylene (PP) material
  • Recyclable after use
By excluding large particles from a sample, the time required to process the mobile phase is shortened. Also, column life is extended by reducing contamination that ultimately causes clogging and back pressure build up.
Applications include: Routine HPLC/UHPLC analysis, composite assays, content uniformity, protein precipitation, solubility testing, dissolution testing and sample filtration. Use with any compound that requires light protection, such as catecholamines or vitamins.
These filters can be used either manually or with a compressor unit. Compressor units allow for processing of multiple samples, further reducing sample preparation time for chromatographic analysis.
PP for general filtration media-/solvent-based samples.
PTFE for aggressive samples, high chemical resistance membrane.
Regenerated cellulose and PVDF for aqueous/organic solvents - low, non specific protein-binding media.
Nylon for aqueous/organic samples in 3 to 10 pH range.
PES for biological samples requiring low protein-binding media.
Depth PP for aqueous/organic solvents - high flow and loading capacity.
GMF for high particulate-laden liquids.
Filtering capacity: 0.4 ml
Maximum operating temperature: 50 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty