Would you like to log in again?
Clear-Rite 3™ Clearant, Epredia
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6901 6905 6901 6905 6915
84000-054EA 317.36 USD InStock
84000-054 CA84000-052 84000-052 CA84000-054 10128-138
Clear-Rite 3™ Clearant, Epredia
Stains Microscopy and staining reagents
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series™ Clear-Rite 3™, a unique blend of isoparaffinic aliphatic hydrocarbons.
  • Very low odor; benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty