Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) alcoholic solution

Supplier: Epredia
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

71204 71204 71211 71225 71225 71211
84000-098EA 739.67 USD InStock
84000-098 CA84000-096 84000-096 CA84000-100 84000-100 CA84000-098
Eosin Y (yellowish) alcoholic solution
Eosin Y (yellowish)
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Epredia™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated Staining Solutions.

  • Strict quality control of raw materials and finished product ensures a high quality product every time
  • Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
  • Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
  • Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y

Epredia™ Eosin-Y Alcoholic is an alcoholic, acidified counterstain. This product will stain cytoplasmic components three distinct color hues. Muscle, red blood cells and connective tissue all stain various shades of red and pink. Epredia™ Eosin-Y Alcoholic will provide a clear contrast between the cytoplasm and nuclei of the cell. No dilution is necessary for this product as it is ready for use. The pH of this Eosin product is between 4.8 and 5.2. Typical staining times are 30-90 seconds.

Epredia™ Eosin-Y w/ Phloxine is an alcoholic, acidified counterstain. Phloxine B has been added to brighten the cytoplasmic stain and give a more intense red color to the connective tissue, especially muscle. Inclusion bodies are better demonstrated by the inclusion of phloxine-B. Some pathologists prefer the look and range of cytoplasmic colors it provides. No dilution is necessary and it is ready for use. (see Cat. No. 84000-102, 84000-104)

Epredia™ Eosin-Y Saturated is an alcohol based, rapid cytoplasmic stain. It does not contain glacial acetic acid, so its pH is much higher (neutral) than other eosin-y alcoholic counterstains. This stain will provide intensities similar to other commercially available eosin-y products. Thermo Scientific Eosin-Y Saturated can be used as is or diluted for use. If the product is diluted, it is recommended to dilute with 70% alcohol to the desired strength. (see Cat. No. 84000-106)


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty