Would you like to log in again?
Differentiation Solution, Epredia
Supplier: Epredia
74211 74211
84000-110EA 182.73 USD InStock
84000-110 CA84000-110
Differentiation Solution, Epredia
Stains Microscopy and staining reagents
Epredia™ Differentiation Solution is a standard formulation of hydrochloric acid in denatured alcohol.
  • Reproducible batch results allow for consistent staining in regressive staining procedures
  • Solution provides clear differentiation and contrast between hematoxylin and eosin
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty