Would you like to log in again?
Papanicolaou's solution EA 50
Supplier: Epredia
75525 75504 75525 75511 75511 75504
84000-120EA 235.19 USD InStock
CA84000-118 84000-120 CA84000-122 84000-122 CA84000-120 84000-118
Papanicolaou's solution EA 50
Papanicolaou's solution EA 50
Control the color hues and intensities of your work with Epredia™ Papanicolaou EA Stain, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Epredia™ EA 36 is used in conjunction with Epredia™ OG 6 for the staining of gynecological specimens. It will provide a slightly lighter cytoplasmic hue than EA 50. EA 36 is recommended for end users who desire a more pastel staining result in gynecological specimens.
IVDLogo
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty