Would you like to log in again?
Richard-Allan Scientific® Saccomanno Fluid, Thermo Scientific
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
76161 76161 76160 76160
84000-146CS 818.75 USD InStock
84000-146 CA84000-148 84000-148 CA84000-146
Richard-Allan Scientific® Saccomanno Fluid, Thermo Scientific
Stains Microscopy and staining reagents
The ready-to-use fixative is a mixture of alcohol and polyethylene glycol that is suited for upper respiratory tract cytology, particularly sputum. The fluid permits storage of specimens for weeks and prevents air drying artifacts from appearing in the samples. It can also serve as a good general cytology fixative. The solution is tinted for easy specimen identification. The poly containers are clear and labeled with a “Do Not Drink” warning, because specimens may be collected at home.
  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
The ready-to-use fixative is a mixture of alcohol and polyethylene glycol that is suited especially for upper respiratory tract cytology, particularly sputum. The fluid permits storage of specimens for weeks and prevents air drying artifacts from appearing in the samples. It can also serve as a good general cytology fixative. The solution is tinted for easy specimen identification. The poly containers are clear and labeled with a 'Do Not Drink' warning, since specimens may be collected at home.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty