Would you like to log in again?
Neg-50™ Frozen Section Medium, Epredia™
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Neg-50™
6506 6502Y 6502G 6506M 6502 6502B 6502P 6506 6502O 6502
84000-154CS 108.77 USD InStock
84000-154 89370-846 89370-848 CA84000-156 89370-842 CA84000-154 89370-844 84000-156 89370-850 89370-852
Neg-50™ Frozen Section Medium, Epredia™
Stains Microscopy and staining reagents
Yield high-quality sections down to–50°C without chatter using Epredia™ Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
This colorless, water-soluble frozen section medium yields high-quality sections down to –50 °C (–58 °F) without chatter. It leaves no residue after staining, and is compatible with the Microm® Automatic Cryo Approach system.
Ordering information: Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty