Would you like to log in again?
Stain Kits and Replacement Components, Epredia
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Richard-Allan Scientific®
87017 88103 87016 88037 88020 88031 88005 88037 88012 88028 88031 87020 88039 87012 88004 88036 88003 88020 88000 88012 87021 88038 87009 87017 87016 87018 88022 87101 87019 88015 87008 88034 88101 88019 87006 88025 88007 88027 88018 88102 87009 87018 88030 88009 88001 88028 88025 88039 88006 87012 88001 87015 87008 88009 88006 87006 88027 88035 88002 88008 88117 88014 88036 87015 88005 88018 88026 88023 87019 88014 88011 87007 88104 88104 87020 88007 88015 87021 88008 88013 87003 88102 88023 88030 88101 88024 88011 88029 88038 88117 88035 88033 88024 87003 88019 88013 88002 88029 87007 88103 88034 88033 88003 88026 88017 88000 88017 88004 88022
84000-242EA 102.31 USD InStock
84000-242 CA84000-250 84000-240 84000-246 84000-244 CA84000-252 CA84000-258 84000-248 CA84000-256 84000-252 CA84000-262 84000-250 CA84000-260 CA84000-266 84000-256 CA84000-264 84000-258 CA84000-268 CA84000-300 CA84000-306 CA84000-304 CA84000-308 84000-260 CA84000-272 84000-264 CA84000-270 84000-262 84000-300 CA84000-310 84000-268 84000-266 CA84000-274 CA84000-314 84000-304 CA84000-312 84000-308 84000-306 84000-270 CA84000-280 84000-274 CA84000-284 84000-272 CA84000-282 CA84000-288 CA84000-200 84000-312 CA84000-286 84000-310 CA84000-204 CA84000-202 84000-314 CA84000-208 CA84000-206 CA84000-290 84000-282 84000-280 CA84000-294 84000-286 CA84000-292 84000-284 CA84000-298 84000-202 CA84000-210 84000-200 CA84000-296 84000-288 84000-206 CA84000-214 CA84000-212 84000-204 CA84000-218 84000-208 CA84000-216 84000-292 84000-290 84000-296 84000-294 CA84000-222 84000-212 CA84000-220 84000-298 84000-210 84000-216 CA84000-226 84000-214 CA84000-224 84000-218 CA84000-228 84000-220 16002-128 CA84000-232 84000-224 84000-222 CA84000-230 84000-228 CA84000-236 CA84000-234 84000-226 CA84000-238 84000-230 CA84000-240 84000-234 CA84000-244 84000-232 CA84000-242 CA84000-248 84000-238 84000-236 CA84000-246
Stain Kits and Replacement Components, Epredia
Stains Histology Stains
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
  • Save time
  • Components available separately
  • All the necessary stains, solutions and components
Acid Fast Bacillus, blue secondary stain kits stain bright red with a pale blue background. Acid Fast Bacillus, green secondary stain kit stains dark red with a pale green background. Demonstrate amyloid with Epredia™ Amyloid Stain. Amyloid stain kit stains orange to red.
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Elastic Stain. Elastic stain kits stain blue-black to black for elastic tissue, red for collagen with a yellow background.
Save time with Epredia™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome. Gomori Trichrome (Blue Collagen) stain kit stains red for Muscle tissue and cytoplasm, blue for collagen and blue-black for nuclei. Epredia™ Gomori Trichrome (Green Collagen is a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Iron Stain. Iron stain kits stain blue iron, pale pink cytoplasm and pink-red nuclei.
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution. Modified Grocott’s Methenamine kit stains black with a green background.
Identify primary tumor sites with the aid of Epredia™ Mucicarmine. Mucicarmine stain kit stains dark rose for mucin and black for nuclei with a yellow background.
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Periodic Acid-Schiff (PAS) Solution. Periodic Acid Schiff kit stains rose to magenta with blue nuclei.
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Steiner-Steiner Silver. Steiner-Steiner silver kits stain black with a yellow to brown background.
Ordering information: All kits come with all stains and solutions necessary to complete the staining procedure. Components of kits are also available separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty